Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258415.1 | NP_001245344.1 | 12 - 13 | 1709 | 139 |
NM_001258416.1 | NP_001245345.1 | 13 - 14 | 1595 | 139 | |
NM_001258417.1 | NP_001245346.1 | 12 - 13 | 1409 | 139 | |
NM_001258418.1 | NP_001245347.1 | 12 - 13 | 1527 | 139 | |
NM_020809.3 | NP_065860.2 | 13 - 14 | 1706 | 139 | |
XM_005271628.3 | XP_005271685.1 | 2 - 3 | 380 | 139 | |
XM_006718893.3 | XP_006718956.1 | 12 - 13 | 1615 | 139 | |
GenBank mRNA | AB037812.1 | - | 12 - 13 | 1431 | 139 |
AK292290.1 | - | 2 - 3 | 380 | 139 | |
AK307602.1 | - | 12 - 13 | 1615 | 139 | |
AY496263.1 | - | 13 - 14 | 1706 | 139 | |
AY496264.1 | - | 12 - 13 | 1409 | 139 | |
AY496265.1 | - | 12 - 13 | 1527 | 139 | |
AY496266.1 | - | 12 - 13 | 1709 | 139 | |
AY496267.1 | - | 13 - 14 | 1595 | 139 | |
BC039340.1 | - | 2 - 3 | 365 | 139 |