Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258415.1 | NP_001245344.1 | 4 - 5 | 795 | 72 |
NM_001258416.1 | NP_001245345.1 | 5 - 6 | 681 | 72 | |
NM_001258417.1 | NP_001245346.1 | 4 - 5 | 495 | 72 | |
NM_001258418.1 | NP_001245347.1 | 4 - 5 | 613 | 72 | |
NM_020809.3 | NP_065860.2 | 5 - 6 | 792 | 72 | |
XM_006718893.3 | XP_006718956.1 | 4 - 5 | 701 | 72 | |
GenBank mRNA | AB037812.1 | - | 4 - 5 | 517 | 72 |
AK307602.1 | - | 4 - 5 | 701 | 72 | |
AY496263.1 | - | 5 - 6 | 792 | 72 | |
AY496264.1 | - | 4 - 5 | 495 | 72 | |
AY496265.1 | - | 4 - 5 | 613 | 72 | |
AY496266.1 | - | 4 - 5 | 795 | 72 | |
AY496267.1 | - | 5 - 6 | 681 | 72 |