Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291880.1 | NP_001278809.1 | 2 - 3 | 1278 | 67 |
NM_004185.4 | NP_004176.2 | 3 - 4 | 1349 | 67 | |
NM_024494.2 | NP_078613.1 | 2 - 3 | 924 | 67 | |
GenBank mRNA | AB045116.1 | - | 3 - 4 | 472 | 67 |
AB045117.1 | - | 2 - 3 | 428 | 67 | |
AK127449.1 | - | 2 - 3 | 1278 | 67 | |
AK312696.1 | - | 2 - 3 | 676 | 67 | |
BC141825.1 | - | 2 - 3 | 924 | 67 |