Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173536.3 | NP_775807.2 | 5 - 6 | 771 | 114 |
XM_017007990.1 | XP_016863479.1 | 3 - 4 | 442 | 114 | |
GenBank mRNA | AK122845.1 | - | 5 - 6 | 787 | 114 |
AK289806.1 | - | 5 - 6 | 769 | 114 | |
BC031087.1 | - | 5 - 6 | 707 | 114 | |
BX648937.1 | - | 5 - 6 | 772 | 114 | |
CR933628.1 | - | 5 - 6 | 705 | 114 |