Assay Design
Gene Transcripts
- Gene Symbol
- HIST1H2BA
- Entrez Gene ID
-
255626
- Gene Name
- histone cluster 1, H2ba
- Gene Aliases
-
H2BFU, STBP, TH2B, TSH2B, TSH2B.1, bA317E16.3
- Chromosome Location
-
Chr.6: 25726909 - 25727345 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK312055.1 |
- |
1 - 1 |
146 |
65 |
View Full Product Details