Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014331.3 | NP_055146.1 | 9 - 10 | 1400 | 68 |
XM_011531802.2 | XP_011530104.1 | 9 - 10 | 1469 | 68 | |
GenBank mRNA | AB026891.1 | - | 9 - 10 | 1355 | 68 |
AB040875.1 | - | 9 - 10 | 1255 | 68 | |
AF200708.1 | - | 9 - 10 | 1365 | 68 | |
AF252872.1 | - | 9 - 10 | 1351 | 68 | |
AJ277882.1 | - | 9 - 10 | 1120 | 68 | |
AK290359.1 | - | 9 - 10 | 1400 | 68 | |
AK314855.1 | - | 9 - 10 | 1294 | 68 | |
BC012087.1 | - | 9 - 10 | 1280 | 68 |