Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025081.1 | NP_001020252.1 | 2 - 3 | 351 | 87 |
NM_002385.2 | NP_002376.1 | 2 - 3 | 351 | 87 | |
GenBank mRNA | AK289893.1 | - | 2 - 3 | 306 | 87 |
AK314553.1 | - | 2 - 3 | 347 | 87 | |
M30515.1 | - | 2 - 3 | 267 | 87 | |
M30516.1 | - | 2 - 3 | 270 | 87 |