Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100122.1 | NP_001093592.1 | 9 - 10 | 1177 | 71 |
NM_014963.2 | NP_055778.2 | 12 - 13 | 1509 | 71 | |
XM_011527804.2 | XP_011526106.1 | 12 - 13 | 1482 | 71 | |
GenBank mRNA | AB023180.1 | - | 12 - 13 | 1486 | 71 |
AK074102.1 | - | 8 - 9 | 969 | 71 | |
AK125139.1 | - | 9 - 10 | 1124 | 71 | |
AK292407.1 | - | 12 - 13 | 1509 | 71 | |
AK296849.1 | - | 9 - 10 | 1124 | 71 | |
AK300981.1 | - | 13 - 14 | 1449 | 71 | |
AK309167.1 | - | 9 - 10 | 1124 | 71 | |
BQ950129.1 | - | 2 - 3 | 268 | 71 |