Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100122.1 | NP_001093592.1 | 20 - 21 | 2608 | 72 |
NM_014963.2 | NP_055778.2 | 23 - 24 | 2940 | 72 | |
XM_011527804.2 | XP_011526106.1 | 23 - 24 | 2913 | 72 | |
GenBank mRNA | AB023180.1 | - | 23 - 24 | 2917 | 72 |
AK074102.1 | - | 19 - 20 | 2400 | 72 | |
AK125139.1 | - | 20 - 21 | 2555 | 72 | |
AK292407.1 | - | 23 - 24 | 2940 | 72 |