Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100122.1 | NP_001093592.1 | 25 - 26 | 3126 | 83 |
NM_014963.2 | NP_055778.2 | 28 - 29 | 3458 | 83 | |
XM_011527804.2 | XP_011526106.1 | 28 - 29 | 3431 | 83 | |
GenBank mRNA | AB023180.1 | - | 28 - 29 | 3435 | 83 |
AK074102.1 | - | 24 - 25 | 2918 | 83 | |
AK125139.1 | - | 25 - 26 | 3073 | 83 | |
AK292407.1 | - | 28 - 29 | 3458 | 83 | |
BC106021.1 | - | 5 - 6 | 530 | 83 |