Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032048.2 | NP_114437.2 | 5 - 6 | 2826 | 110 |
GenBank mRNA | AB026706.1 | - | 1 - 2 | 132 | 110 |
AF270513.1 | - | 5 - 6 | 2826 | 110 | |
AK090519.1 | - | 2 - 3 | 921 | 110 | |
BC136541.1 | - | 5 - 6 | 2780 | 110 |