Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032048.2 | NP_114437.2 | 7 - 8 | 2982 | 118 |
GenBank mRNA | AB026706.1 | - | 3 - 4 | 288 | 118 |
AF270513.1 | - | 7 - 8 | 2982 | 118 | |
AK090519.1 | - | 4 - 5 | 1077 | 118 | |
BC032231.1 | - | 5 - 6 | 655 | 118 | |
BC136541.1 | - | 7 - 8 | 2936 | 118 |