Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171137.1 | NP_001164608.1 | 13 - 14 | 1751 | 72 |
NM_018387.4 | NP_060857.2 | 13 - 14 | 1768 | 72 | |
NR_033234.1 | 13 - 14 | 1693 | 72 | ||
XM_017014890.1 | XP_016870379.1 | 12 - 13 | 1595 | 72 | |
XM_017014891.1 | XP_016870380.1 | 12 - 13 | 1568 | 72 | |
XM_017014892.1 | XP_016870381.1 | 12 - 13 | 1827 | 72 | |
XM_017014893.1 | XP_016870382.1 | 12 - 13 | 1537 | 72 | |
XM_017014894.1 | XP_016870383.1 | 13 - 14 | 3139 | 72 | |
XM_017014895.1 | XP_016870384.1 | 13 - 14 | 3136 | 72 | |
XM_017014896.1 | XP_016870385.1 | 13 - 14 | 1956 | 72 | |
XM_017014897.1 | XP_016870386.1 | 12 - 13 | 1595 | 72 | |
XM_017014898.1 | XP_016870387.1 | 12 - 13 | 1595 | 72 | |
GenBank mRNA | AF333337.1 | - | 11 - 12 | 1368 | 72 |
AK002169.1 | - | 7 - 8 | 795 | 72 | |
AK024285.1 | - | 5 - 6 | 606 | 72 | |
AK027890.1 | - | 12 - 13 | 1685 | 72 | |
AL136866.1 | - | 13 - 14 | 1751 | 72 | |
AL833947.1 | - | 13 - 14 | 1693 | 72 | |
AL834150.1 | - | 13 - 14 | 1693 | 72 | |
BC002693.2 | - | 10 - 11 | 1167 | 72 | |
BC017732.1 | - | 13 - 14 | 1768 | 72 | |
BC108731.1 | - | 12 - 13 | 1660 | 72 | |
EU794683.1 | - | 13 - 14 | 1768 | 72 | |
KU178696.1 | - | 10 - 11 | 1338 | 72 | |
KU178697.1 | - | 10 - 11 | 1338 | 72 |