Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171137.1 | NP_001164608.1 | 15 - 16 | 2041 | 66 |
NM_018387.4 | NP_060857.2 | 15 - 16 | 2058 | 66 | |
NR_033234.1 | 15 - 16 | 1983 | 66 | ||
XM_017014890.1 | XP_016870379.1 | 14 - 15 | 1885 | 66 | |
XM_017014891.1 | XP_016870380.1 | 14 - 15 | 1858 | 66 | |
XM_017014892.1 | XP_016870381.1 | 14 - 15 | 2117 | 66 | |
XM_017014893.1 | XP_016870382.1 | 14 - 15 | 1827 | 66 | |
XM_017014894.1 | XP_016870383.1 | 15 - 16 | 3429 | 66 | |
XM_017014895.1 | XP_016870384.1 | 15 - 16 | 3426 | 66 | |
XM_017014896.1 | XP_016870385.1 | 15 - 16 | 2246 | 66 | |
XM_017014897.1 | XP_016870386.1 | 14 - 15 | 1885 | 66 | |
XM_017014898.1 | XP_016870387.1 | 14 - 15 | 1885 | 66 | |
GenBank mRNA | AF333337.1 | - | 13 - 14 | 1658 | 66 |
AK002169.1 | - | 9 - 10 | 1085 | 66 | |
AK024285.1 | - | 7 - 8 | 896 | 66 | |
AK027890.1 | - | 14 - 15 | 1975 | 66 | |
AL136866.1 | - | 15 - 16 | 2041 | 66 | |
AL833947.1 | - | 15 - 16 | 1983 | 66 | |
AL834150.1 | - | 15 - 16 | 1983 | 66 | |
BC002693.2 | - | 12 - 13 | 1457 | 66 | |
BC017732.1 | - | 15 - 16 | 2058 | 66 | |
BC108731.1 | - | 14 - 15 | 1950 | 66 | |
EU794683.1 | - | 15 - 16 | 2058 | 66 | |
KU178696.1 | - | 12 - 13 | 1628 | 66 | |
KU178697.1 | - | 12 - 13 | 1628 | 66 |