Assay Design



Gene Transcripts

Gene Symbol
COQ8A
Entrez Gene ID
56997
Gene Name
coenzyme Q8A
Gene Aliases
ADCK3, ARCA2, CABC1, COQ10D4, COQ8, SCAR9
Chromosome Location
Chr.1: 226939339 - 226987545 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020247.4 NP_064632.2 14 - 15 1830 128
XM_005273201.1 XP_005273258.1 14 - 15 1726 128
XM_011544238.1 XP_011542540.1 15 - 16 1922 128
XM_011544239.2 XP_011542541.1 16 - 17 1945 128
XM_011544240.2 XP_011542542.1 14 - 15 1739 128
XM_011544241.2 XP_011542543.1 14 - 15 1701 128
XM_017001852.1 XP_016857341.1 14 - 15 1920 128
GenBank mRNA AB073905.1 - 14 - 15 1753 128
AF218003.1 - 6 - 7 845 128
AJ278126.1 - 10 - 11 1048 128
AK074693.1 - 14 - 15 1773 128
AK090494.1 - 14 - 15 1567 128
AK092784.1 - 14 - 15 1553 128
AK293571.1 - 11 - 12 1193 128
AK297735.1 - 11 - 12 1109 128
AK300537.1 - 10 - 11 919 128
AK301012.1 - 16 - 17 1751 128
AL137511.1 - 1 - 2 128
BC005171.2 - 14 - 15 1779 128
BX648860.1 - 14 - 15 1651 128

View Full Product Details