Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006716795.2 | XP_006716858.1 | 2 - 3 | 270 | 67 |
XM_006716798.3 | XP_006716861.1 | 3 - 4 | 275 | 67 | |
XM_006716799.3 | XP_006716862.1 | 2 - 3 | 377 | 67 | |
XM_006716800.2 | XP_006716863.1 | 2 - 3 | 225 | 67 | |
XM_011517952.1 | XP_011516254.1 | 2 - 3 | 213 | 67 | |
XM_011517956.1 | XP_011516258.1 | 2 - 3 | 270 | 67 | |
XM_011517958.1 | XP_011516260.1 | 1 - 2 | 131 | 67 | |
XM_017014881.1 | XP_016870370.1 | 2 - 3 | 270 | 67 | |
XM_017014882.1 | XP_016870371.1 | 2 - 3 | 270 | 67 | |
XM_017014883.1 | XP_016870372.1 | 2 - 3 | 270 | 67 | |
XM_017014884.1 | XP_016870373.1 | 2 - 3 | 225 | 67 | |
XM_017014885.1 | XP_016870374.1 | 2 - 3 | 270 | 67 | |
GenBank mRNA | AK000920.1 | - | 3 - 4 | 251 | 67 |
AK301790.1 | - | 2 - 3 | 370 | 67 | |
BC034293.1 | - | 2 - 3 | 344 | 67 | |
BC036349.1 | - | 3 - 4 | 269 | 67 | |
BX649030.1 | - | 2 - 3 | 320 | 67 |