Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006716795.2 | XP_006716858.1 | 5 - 6 | 491 | 80 |
XM_006716798.3 | XP_006716861.1 | 6 - 7 | 496 | 80 | |
XM_006716799.3 | XP_006716862.1 | 4 - 5 | 556 | 80 | |
XM_006716800.2 | XP_006716863.1 | 5 - 6 | 446 | 80 | |
XM_006716801.2 | XP_006716864.1 | 4 - 5 | 547 | 80 | |
XM_011517952.1 | XP_011516254.1 | 5 - 6 | 434 | 80 | |
XM_011517954.1 | XP_011516256.1 | 3 - 4 | 487 | 80 | |
XM_011517956.1 | XP_011516258.1 | 5 - 6 | 491 | 80 | |
XM_011517957.1 | XP_011516259.1 | 3 - 4 | 487 | 80 | |
XM_011517958.1 | XP_011516260.1 | 3 - 4 | 310 | 80 | |
XM_017014881.1 | XP_016870370.1 | 5 - 6 | 491 | 80 | |
XM_017014882.1 | XP_016870371.1 | 5 - 6 | 491 | 80 | |
XM_017014883.1 | XP_016870372.1 | 5 - 6 | 491 | 80 | |
XM_017014884.1 | XP_016870373.1 | 4 - 5 | 404 | 80 | |
XM_017014885.1 | XP_016870374.1 | 5 - 6 | 491 | 80 | |
XM_017014886.1 | XP_016870375.1 | 3 - 4 | 303 | 80 | |
XM_017014887.1 | XP_016870376.1 | 3 - 4 | 385 | 80 | |
GenBank mRNA | AK000920.1 | - | 5 - 6 | 430 | 80 |
AK301790.1 | - | 5 - 6 | 591 | 80 | |
BC034293.1 | - | 4 - 5 | 523 | 80 | |
BC036349.1 | - | 6 - 7 | 490 | 80 | |
BX649030.1 | - | 5 - 6 | 541 | 80 |