Assay Design



Gene Transcripts

Gene Symbol
FGA
Entrez Gene ID
2243
Gene Name
fibrinogen alpha chain
Gene Aliases
Fib2
Chromosome Location
Chr.4: 154583126 - 154590766 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000508.4 NP_000499.1 2 - 3 266 60
NM_021871.3 NP_068657.1 2 - 3 266 60
GenBank mRNA AK290559.1 - 2 - 3 250 60
BC070246.1 - 2 - 3 245 60
BC098280.1 - 2 - 3 210 60
BC099720.1 - 2 - 3 210 60
BC101935.1 - 2 - 3 210 60
BC105803.1 - 2 - 3 249 60
BG565064.1 - 2 - 3 246 60
BX494953.1 - 2 - 3 245 60
CN393696.1 - 2 - 3 283 60
J00127.1 - 2 - 3 217 60
J00128.1 - 2 - 3 241 60
KU178062.1 - 2 - 3 187 60
KU178063.1 - 2 - 3 187 60
KU178064.1 - 2 - 3 187 60
M58569.1 - 2 - 3 216 60

View Full Product Details