Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109.4 | NP_001100.3 | 22 - 23 | 2481 | 127 |
NM_001164489.1 | NP_001157961.1 | 21 - 22 | 2314 | 127 | |
XM_017015465.1 | XP_016870954.1 | 23 - 24 | 2662 | 127 | |
GenBank mRNA | AK312834.1 | - | 22 - 23 | 2446 | 127 |
BC064500.1 | - | 21 - 22 | 2261 | 127 | |
BC115404.1 | - | 22 - 23 | 2400 | 127 | |
CR997306.1 | - | 3 - 4 | 340 | 127 | |
D26579.1 | - | 22 - 23 | 2404 | 127 |