Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109.4 | NP_001100.3 | 5 - 6 | 462 | 104 |
NM_001164489.1 | NP_001157961.1 | 5 - 6 | 462 | 104 | |
XM_011539117.2 | XP_011537419.2 | 6 - 7 | 643 | 104 | |
XM_017015465.1 | XP_016870954.1 | 6 - 7 | 643 | 104 | |
XM_017015466.1 | XP_016870955.1 | 6 - 7 | 645 | 104 | |
GenBank mRNA | AK304324.1 | - | 5 - 6 | 619 | 104 |
AK310662.1 | - | 4 - 5 | 318 | 104 | |
AK312834.1 | - | 5 - 6 | 427 | 104 | |
BC064500.1 | - | 5 - 6 | 409 | 104 | |
BC115404.1 | - | 5 - 6 | 381 | 104 | |
DB142906.1 | - | 4 - 5 | 342 | 104 | |
DC401057.1 | - | 5 - 6 | 463 | 104 |