Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000028.2 | NP_000019.2 | 18 - 19 | 2917 | 90 |
NM_000642.2 | NP_000633.2 | 18 - 19 | 2839 | 90 | |
NM_000643.2 | NP_000634.2 | 18 - 19 | 2637 | 90 | |
NM_000644.2 | NP_000635.2 | 18 - 19 | 2577 | 90 | |
NM_000645.2 | NP_000636.2 | 16 - 17 | 2650 | 90 | |
NM_000646.2 | NP_000637.2 | 18 - 19 | 2650 | 90 | |
XM_005270557.2 | XP_005270614.1 | 18 - 19 | 2532 | 90 | |
XM_017000501.1 | XP_016855990.1 | 6 - 7 | 766 | 90 | |
GenBank mRNA | AB208867.1 | - | 17 - 18 | 2443 | 90 |
AB621804.1 | - | 18 - 19 | 2787 | 90 | |
BC078663.1 | - | 18 - 19 | 2559 | 90 | |
M85168.1 | - | 16 - 17 | 2652 | 90 | |
U84007.1 | - | 18 - 19 | 2839 | 90 | |
U84008.1 | - | 18 - 19 | 2577 | 90 | |
U84009.1 | - | 18 - 19 | 2637 | 90 | |
U84010.1 | - | 18 - 19 | 2917 | 90 | |
U84011.1 | - | 18 - 19 | 2650 | 90 |