Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000028.2 | NP_000019.2 | 25 - 26 | 3833 | 141 |
NM_000642.2 | NP_000633.2 | 25 - 26 | 3755 | 141 | |
NM_000643.2 | NP_000634.2 | 25 - 26 | 3553 | 141 | |
NM_000644.2 | NP_000635.2 | 25 - 26 | 3493 | 141 | |
NM_000645.2 | NP_000636.2 | 23 - 24 | 3566 | 141 | |
NM_000646.2 | NP_000637.2 | 25 - 26 | 3566 | 141 | |
XM_005270557.2 | XP_005270614.1 | 25 - 26 | 3448 | 141 | |
XM_017000501.1 | XP_016855990.1 | 13 - 14 | 1682 | 141 | |
GenBank mRNA | AB208867.1 | - | 24 - 25 | 3359 | 141 |
AB621804.1 | - | 25 - 26 | 3703 | 141 | |
BC078663.1 | - | 25 - 26 | 3475 | 141 | |
M85168.1 | - | 23 - 24 | 3568 | 141 | |
U84007.1 | - | 25 - 26 | 3755 | 141 | |
U84008.1 | - | 25 - 26 | 3493 | 141 | |
U84009.1 | - | 25 - 26 | 3553 | 141 | |
U84010.1 | - | 25 - 26 | 3833 | 141 | |
U84011.1 | - | 25 - 26 | 3566 | 141 |