Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000028.2 | NP_000019.2 | 3 - 4 | 777 | 112 |
NM_000642.2 | NP_000633.2 | 3 - 4 | 699 | 112 | |
NM_000643.2 | NP_000634.2 | 3 - 4 | 497 | 112 | |
NM_000644.2 | NP_000635.2 | 3 - 4 | 437 | 112 | |
NM_000645.2 | NP_000636.2 | 1 - 2 | 510 | 112 | |
NM_000646.2 | NP_000637.2 | 3 - 4 | 510 | 112 | |
XM_005270557.2 | XP_005270614.1 | 3 - 4 | 392 | 112 | |
GenBank mRNA | AB208867.1 | - | 2 - 3 | 303 | 112 |
AB621804.1 | - | 3 - 4 | 647 | 112 | |
BC020705.1 | - | 3 - 4 | 619 | 112 | |
BC057220.1 | - | 3 - 4 | 623 | 112 | |
BC078663.1 | - | 3 - 4 | 419 | 112 | |
M85168.1 | - | 1 - 2 | 512 | 112 | |
U84007.1 | - | 3 - 4 | 699 | 112 | |
U84008.1 | - | 3 - 4 | 437 | 112 | |
U84009.1 | - | 3 - 4 | 497 | 112 | |
U84010.1 | - | 3 - 4 | 777 | 112 | |
U84011.1 | - | 3 - 4 | 510 | 112 |