Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000028.2 | NP_000019.2 | 8 - 9 | 1562 | 161 |
NM_000642.2 | NP_000633.2 | 8 - 9 | 1484 | 161 | |
NM_000643.2 | NP_000634.2 | 8 - 9 | 1282 | 161 | |
NM_000644.2 | NP_000635.2 | 8 - 9 | 1222 | 161 | |
NM_000645.2 | NP_000636.2 | 6 - 7 | 1295 | 161 | |
NM_000646.2 | NP_000637.2 | 8 - 9 | 1295 | 161 | |
XM_005270557.2 | XP_005270614.1 | 8 - 9 | 1177 | 161 | |
GenBank mRNA | AB208867.1 | - | 7 - 8 | 1088 | 161 |
AB621804.1 | - | 8 - 9 | 1432 | 161 | |
BC078663.1 | - | 8 - 9 | 1204 | 161 | |
M85168.1 | - | 6 - 7 | 1297 | 161 | |
U84007.1 | - | 8 - 9 | 1484 | 161 | |
U84008.1 | - | 8 - 9 | 1222 | 161 | |
U84009.1 | - | 8 - 9 | 1282 | 161 | |
U84010.1 | - | 8 - 9 | 1562 | 161 | |
U84011.1 | - | 8 - 9 | 1295 | 161 |