Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272042.1 | NP_001258971.1 | 2 - 3 | 334 | 135 |
NM_001272043.1 | NP_001258972.1 | 2 - 3 | 334 | 135 | |
NM_001272044.1 | NP_001258973.1 | 3 - 4 | 463 | 264 | |
NM_001272060.1 | NP_001258989.1 | 2 - 3 | 334 | 135 | |
NM_003161.3 | NP_003152.1 | 2 - 3 | 334 | 135 | |
XM_011525101.2 | XP_011523403.1 | 2 - 3 | 329 | 135 | |
XM_011525102.2 | XP_011523404.1 | 2 - 3 | 329 | 135 | |
XM_011525103.2 | XP_011523405.1 | 3 - 4 | 383 | 264 | |
XM_017024929.1 | XP_016880418.1 | 3 - 4 | 392 | 264 | |
XM_017024930.1 | XP_016880419.1 | 2 - 3 | 293 | 135 | |
XM_017024931.1 | XP_016880420.1 | 2 - 3 | 254 | 135 | |
XM_017024932.1 | XP_016880421.1 | 2 - 3 | 254 | 135 | |
XM_017024933.1 | XP_016880422.1 | 2 - 3 | 249 | 135 | |
GenBank mRNA | AK297147.1 | - | 3 - 4 | 270 | 264 |
AK300202.1 | - | 2 - 3 | 250 | 135 | |
AK312875.1 | - | 2 - 3 | 240 | 135 | |
BC036033.1 | - | 2 - 3 | 294 | 135 | |
BC053365.1 | - | 2 - 3 | 236 | 135 | |
BP359158.1 | - | 2 - 3 | 334 | 135 | |
M60724.1 | - | 2 - 3 | 222 | 135 |