Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000373.3 | NP_000364.1 | 5 - 6 | 1372 | 153 |
NR_033434.1 | 4 - 5 | 1218 | 153 | ||
NR_033437.1 | 6 - 7 | 1471 | 153 | ||
GenBank mRNA | AB062285.1 | - | 5 - 6 | 1294 | 153 |
AK291306.1 | - | 5 - 6 | 1288 | 153 | |
BC000364.2 | - | 5 - 6 | 1289 | 153 | |
BC007511.2 | - | 5 - 6 | 1289 | 153 | |
CR456787.1 | - | 5 - 6 | 1266 | 153 | |
D86227.1 | - | 5 - 6 | 1285 | 153 | |
D86228.1 | - | 5 - 6 | 1285 | 153 | |
D86230.1 | - | 5 - 6 | 1285 | 153 | |
EU921886.1 | - | 5 - 6 | 1286 | 153 | |
EU921887.1 | - | 4 - 5 | 1132 | 153 | |
EU921888.1 | - | 4 - 5 | 1132 | 153 | |
EU921889.1 | - | 5 - 6 | 1231 | 153 | |
EU921890.1 | - | 5 - 6 | 1490 | 153 | |
EU921891.1 | - | 6 - 7 | 1385 | 153 | |
EU921892.1 | - | 5 - 6 | 968 | 153 | |
EU921893.1 | - | 5 - 6 | 939 | 153 | |
EU921894.1 | - | 5 - 6 | 1286 | 153 | |
J03626.1 | - | 5 - 6 | 1370 | 153 | |
KU178375.1 | - | 5 - 6 | 1266 | 153 | |
KU178376.1 | - | 6 - 7 | 1365 | 153 | |
M36661.1 | - | 5 - 6 | 1232 | 153 |