Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083893.1 | NP_001077362.1 | 12 - 13 | 1818 | 147 |
NM_014574.3 | NP_055389.3 | 10 - 11 | 1566 | 147 | |
XM_005267569.3 | XP_005267626.1 | 11 - 12 | 1834 | 147 | |
XM_005267570.3 | XP_005267627.1 | 11 - 12 | 1804 | 147 | |
GenBank mRNA | AF243424.1 | - | 11 - 12 | 1247 | 147 |
AK290229.1 | - | 10 - 11 | 1544 | 147 | |
AK314041.1 | - | 10 - 11 | 1532 | 147 | |
BC126221.1 | - | 10 - 11 | 1484 | 147 | |
BC132673.1 | - | 10 - 11 | 1484 | 147 | |
BC143933.1 | - | 9 - 10 | 1350 | 147 | |
BM453981.1 | - | 4 - 5 | 460 | 147 | |
U17989.1 | - | 10 - 11 | 1554 | 147 |