Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083893.1 | NP_001077362.1 | 6 - 7 | 1052 | 82 |
NM_014574.3 | NP_055389.3 | 6 - 7 | 1052 | 82 | |
XM_005267569.3 | XP_005267626.1 | 6 - 7 | 1179 | 82 | |
XM_005267570.3 | XP_005267627.1 | 6 - 7 | 1179 | 82 | |
GenBank mRNA | AF243424.1 | - | 5 - 6 | 481 | 82 |
AK290229.1 | - | 6 - 7 | 1030 | 82 | |
AK314041.1 | - | 6 - 7 | 1018 | 82 | |
BC126221.1 | - | 6 - 7 | 970 | 82 | |
BC132673.1 | - | 6 - 7 | 970 | 82 | |
BC143933.1 | - | 6 - 7 | 970 | 82 | |
U17989.1 | - | 6 - 7 | 1040 | 82 |