Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304349.1 | NP_001291278.1 | 3 - 4 | 870 | 67 |
NM_002730.3 | NP_002721.1 | 3 - 4 | 436 | 67 | |
NM_207518.2 | NP_997401.1 | 3 - 4 | 245 | 67 | |
XM_017026948.1 | XP_016882437.1 | 3 - 4 | 292 | 67 | |
GenBank mRNA | AF208004.1 | - | 3 - 4 | 230 | 67 |
AF239744.1 | - | 3 - 4 | 375 | 67 | |
AK131561.1 | - | 3 - 4 | 870 | 67 | |
AK290147.1 | - | 4 - 5 | 318 | 67 | |
AK292284.1 | - | 3 - 4 | 245 | 67 | |
AK301264.1 | - | 3 - 4 | 605 | 67 | |
BC039846.1 | - | 3 - 4 | 432 | 67 | |
BC108259.1 | - | 3 - 4 | 433 | 67 | |
BU543573.1 | - | - | 436 | 67 | |
M80335.1 | - | 2 - 3 | 185 | 67 | |
X07767.1 | - | 3 - 4 | 316 | 67 |