Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013438.4 | NP_038466.2 | 6 - 7 | 1676 | 67 |
NM_053067.2 | NP_444295.1 | 6 - 7 | 1676 | 67 | |
XM_005251948.2 | XP_005252005.1 | 6 - 7 | 1406 | 67 | |
GenBank mRNA | AB035275.1 | - | 6 - 7 | 1272 | 67 |
AB209468.1 | - | 5 - 6 | 820 | 67 | |
AF176069.3 | - | 6 - 7 | 1102 | 67 | |
AF293384.1 | - | 6 - 7 | 1102 | 67 | |
AK027556.1 | - | 6 - 7 | 1375 | 67 | |
AK027576.1 | - | 6 - 7 | 1366 | 67 | |
AL136643.1 | - | 6 - 7 | 1380 | 67 | |
BC010066.2 | - | 6 - 7 | 1356 | 67 | |
BC039294.1 | - | 6 - 7 | 1367 | 67 |