Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019106.5 | NP_061979.3 | 8 - 9 | 1004 | 70 |
NM_145733.2 | NP_663786.2 | 8 - 9 | 1004 | 70 | |
XM_017028862.1 | XP_016884351.1 | 9 - 10 | 2602 | 70 | |
GenBank mRNA | AB209152.1 | - | 5 - 6 | 471 | 70 |
AF285107.1 | - | 8 - 9 | 881 | 70 | |
AF285108.1 | - | 6 - 7 | 586 | 70 | |
AF285109.1 | - | 8 - 9 | 881 | 70 | |
AK290037.1 | - | 8 - 9 | 989 | 70 | |
AK299919.1 | - | 7 - 8 | 749 | 70 | |
AL833942.1 | - | 8 - 9 | 939 | 70 | |
BC111779.1 | - | 8 - 9 | 897 | 70 | |
CR456572.1 | - | 8 - 9 | 893 | 70 |