Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165927.1 | NP_001159399.1 | 13 - 14 | 1228 | 101 |
NM_001321268.1 | NP_001308197.1 | 12 - 13 | 1141 | 101 | |
NM_001321269.1 | NP_001308198.1 | 13 - 14 | 1239 | 101 | |
NM_017777.3 | NP_060247.2 | 13 - 14 | 1239 | 101 | |
XM_005257485.3 | XP_005257542.1 | 13 - 14 | 1220 | 101 | |
XM_006721965.2 | XP_006722028.1 | 12 - 13 | 1073 | 101 | |
XM_011524957.2 | XP_011523259.1 | 13 - 14 | 1219 | 101 | |
XM_011524958.2 | XP_011523260.1 | 13 - 14 | 1219 | 101 | |
XM_011524959.2 | XP_011523261.1 | 13 - 14 | 1218 | 101 | |
XM_011524960.2 | XP_011523262.1 | 13 - 14 | 1218 | 101 | |
XM_017024803.1 | XP_016880292.1 | 13 - 14 | 1187 | 101 | |
XM_017024804.1 | XP_016880293.1 | 13 - 14 | 1187 | 101 | |
XM_017024805.1 | XP_016880294.1 | 13 - 14 | 1253 | 101 | |
GenBank mRNA | AK000352.1 | - | 10 - 11 | 909 | 101 |
AK301020.1 | - | 12 - 13 | 1072 | 101 | |
AK310815.1 | - | 13 - 14 | 1228 | 101 | |
BC010061.2 | - | 13 - 14 | 1174 | 101 | |
CR457229.1 | - | 9 - 10 | 735 | 101 | |
DQ185029.1 | - | 13 - 14 | 1239 | 101 | |
KU178661.1 | - | 9 - 10 | 735 | 101 | |
KU178662.1 | - | 9 - 10 | 735 | 101 | |
KU178663.1 | - | 9 - 10 | 735 | 101 |