Assay Design



Gene Transcripts

Gene Symbol
COQ8A
Entrez Gene ID
56997
Gene Name
coenzyme Q8A
Gene Aliases
ADCK3, ARCA2, CABC1, COQ10D4, COQ8, SCAR9
Chromosome Location
Chr.1: 226939339 - 226987545 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017001852.1 XP_016857341.1 1 - 2 252 124
GenBank mRNA BC129930.1 - 7 - 8 1004 124

View Full Product Details