Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301627.1 | NP_001288556.1 | 7 - 8 | 1259 | 133 |
NM_052917.3 | NP_443149.2 | 9 - 10 | 1737 | 133 | |
XM_011510537.2 | XP_011508839.1 | 9 - 10 | 1732 | 133 | |
XM_011510538.2 | XP_011508840.1 | 7 - 8 | 1144 | 133 | |
XM_017003258.1 | XP_016858747.1 | 9 - 10 | 1586 | 133 | |
XM_017003259.1 | XP_016858748.1 | 8 - 9 | 1311 | 133 | |
XM_017003260.1 | XP_016858749.1 | 9 - 10 | 1732 | 133 | |
XM_017003261.1 | XP_016858750.1 | 9 - 10 | 1596 | 133 | |
XM_017003262.1 | XP_016858751.1 | 7 - 8 | 1002 | 133 | |
XM_017003263.1 | XP_016858752.1 | 7 - 8 | 1144 | 133 | |
GenBank mRNA | AB067505.1 | - | 7 - 8 | 1037 | 133 |
AB078142.1 | - | 7 - 8 | 1155 | 133 | |
AJ505991.2 | - | 7 - 8 | 1155 | 133 | |
AK123152.1 | - | - | 1420 | 133 | |
AK131195.1 | - | 9 - 10 | 1682 | 133 | |
AK308675.1 | - | 7 - 8 | 1144 | 133 | |
BC101031.1 | - | 7 - 8 | 1155 | 133 | |
BC101032.2 | - | 7 - 8 | 1155 | 133 | |
BC101033.1 | - | 7 - 8 | 1155 | 133 | |
BC101034.2 | - | 7 - 8 | 1155 | 133 | |
KJ534843.1 | - | 7 - 8 | 1155 | 133 | |
KJ535058.1 | - | 6 - 7 | 986 | 133 |