Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001185075.1 | NP_001172004.1 | 10 - 11 | 1216 | 101 |
NM_001185076.1 | NP_001172005.1 | 10 - 11 | 1216 | 101 | |
NM_001185081.1 | NP_001172010.1 | 10 - 11 | 1216 | 101 | |
NM_001185082.1 | NP_001172011.1 | 10 - 11 | 1216 | 101 | |
NM_002024.5 | NP_002015.1 | 10 - 11 | 1216 | 101 | |
NR_033699.1 | 10 - 11 | 1216 | 101 | ||
NR_033700.1 | 10 - 11 | 1216 | 101 | ||
GenBank mRNA | AB209188.1 | - | 6 - 7 | 1706 | 101 |
AK302707.1 | - | 5 - 6 | 540 | 101 | |
BC086957.1 | - | 10 - 11 | 1246 | 101 | |
BQ287827.1 | - | 1 - 2 | 101 | ||
BX498094.1 | - | 1 - 2 | 54 | 101 | |
CN315330.1 | - | 1 - 2 | 90 | 101 | |
KC774027.1 | - | 10 - 11 | 1049 | 101 | |
KC774028.1 | - | 10 - 11 | 1049 | 101 | |
KJ534836.1 | - | 10 - 11 | 987 | 101 | |
KJ534837.1 | - | 10 - 11 | 987 | 101 | |
KJ534838.1 | - | 10 - 11 | 987 | 101 | |
KM091457.1 | - | 11 - 12 | 1095 | 101 | |
KM091458.1 | - | 11 - 12 | 1095 | 101 | |
KM091459.1 | - | 11 - 12 | 1095 | 101 | |
M67468.1 | - | 10 - 11 | 1176 | 101 | |
S65791.1 | - | 10 - 11 | 1206 | 101 | |
X69962.1 | - | 10 - 11 | 1206 | 101 |