Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159285.1 | NP_001152757.1 | 6 - 7 | 878 | 76 |
NM_001159295.1 | NP_001152767.1 | 6 - 7 | 942 | 76 | |
NM_139243.3 | NP_640336.1 | 7 - 8 | 956 | 76 | |
XM_005262741.2 | XP_005262798.1 | 6 - 7 | 977 | 76 | |
XM_005262743.2 | XP_005262800.1 | 6 - 7 | 877 | 76 | |
XM_005262744.2 | XP_005262801.1 | 6 - 7 | 977 | 76 | |
XM_005262745.2 | XP_005262802.1 | 5 - 6 | 908 | 76 | |
XM_011531603.1 | XP_011529905.1 | 6 - 7 | 875 | 76 | |
GenBank mRNA | AK057303.1 | - | 7 - 8 | 910 | 76 |
AK093297.1 | - | 6 - 7 | 866 | 76 | |
BC040229.1 | - | 6 - 7 | 824 | 76 |