Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159295.1 | NP_001152767.1 | 7 - 8 | 1062 | 96 |
NM_139243.3 | NP_640336.1 | 8 - 9 | 1076 | 96 | |
XM_005262741.2 | XP_005262798.1 | 7 - 8 | 1097 | 96 | |
XM_005262743.2 | XP_005262800.1 | 7 - 8 | 997 | 96 | |
XM_005262745.2 | XP_005262802.1 | 6 - 7 | 1028 | 96 | |
XM_011531603.1 | XP_011529905.1 | 7 - 8 | 995 | 96 | |
GenBank mRNA | AK057303.1 | - | 8 - 9 | 1030 | 96 |
AK093297.1 | - | 7 - 8 | 986 | 96 |