Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159285.1 | NP_001152757.1 | 8 - 9 | 1144 | 87 |
NM_001159295.1 | NP_001152767.1 | 8 - 9 | 1241 | 87 | |
NM_139243.3 | NP_640336.1 | 9 - 10 | 1255 | 87 | |
XM_005262741.2 | XP_005262798.1 | 8 - 9 | 1276 | 87 | |
XM_005262743.2 | XP_005262800.1 | 8 - 9 | 1176 | 87 | |
XM_005262744.2 | XP_005262801.1 | 8 - 9 | 1243 | 87 | |
XM_005262745.2 | XP_005262802.1 | 7 - 8 | 1207 | 87 | |
XM_011531603.1 | XP_011529905.1 | 8 - 9 | 1174 | 87 | |
GenBank mRNA | AK057303.1 | - | 9 - 10 | 1209 | 87 |
AK093297.1 | - | 8 - 9 | 1165 | 87 | |
BC040229.1 | - | 8 - 9 | 1090 | 87 |