Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159285.1 | NP_001152757.1 | 9 - 10 | 1372 | 96 |
NM_001159295.1 | NP_001152767.1 | 9 - 10 | 1469 | 96 | |
NM_139243.3 | NP_640336.1 | 10 - 11 | 1483 | 96 | |
XM_005262741.2 | XP_005262798.1 | 9 - 10 | 1504 | 96 | |
XM_005262743.2 | XP_005262800.1 | 9 - 10 | 1404 | 96 | |
XM_005262744.2 | XP_005262801.1 | 9 - 10 | 1471 | 96 | |
XM_005262745.2 | XP_005262802.1 | 8 - 9 | 1435 | 96 | |
XM_011531603.1 | XP_011529905.1 | 9 - 10 | 1402 | 96 | |
GenBank mRNA | AK057303.1 | - | 10 - 11 | 1437 | 96 |
AK093297.1 | - | 9 - 10 | 1393 | 96 | |
BC040229.1 | - | 9 - 10 | 1318 | 96 |