Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003809.2 | NP_001003809.1 | 9 - 10 | 1888 | 142 |
NM_001242762.1 | NP_001229691.1 | 9 - 10 | 1831 | 142 | |
NM_001242763.1 | NP_001229692.1 | 10 - 11 | 1945 | 142 | |
NM_001242764.1 | NP_001229693.1 | 9 - 10 | 1861 | 142 | |
NM_001242766.1 | NP_001229695.1 | 10 - 11 | 1918 | 142 | |
NM_001308390.1 | NP_001295319.1 | 10 - 11 | 1943 | 142 | |
NM_004746.3 | NP_004737.2 | 12 - 13 | 3245 | 142 | |
XM_005258171.2 | XP_005258228.1 | 13 - 14 | 3316 | 142 | |
XM_005258172.1 | XP_005258229.1 | 13 - 14 | 3311 | 142 | |
XM_005258173.3 | XP_005258230.2 | 11 - 12 | 2107 | 142 | |
XM_005258174.4 | XP_005258231.1 | 10 - 11 | 2164 | 142 | |
XM_006722367.3 | XP_006722430.1 | 11 - 12 | 2275 | 142 | |
XM_017026080.1 | XP_016881569.1 | 10 - 11 | 2244 | 142 | |
XM_017026081.1 | XP_016881570.1 | 9 - 10 | 1908 | 142 | |
XM_017026083.1 | XP_016881572.1 | 9 - 10 | 2133 | 142 | |
GenBank mRNA | AB000276.1 | - | 9 - 10 | 1831 | 142 |
AB000277.1 | - | 9 - 10 | 2785 | 142 | |
AK294717.1 | - | 9 - 10 | 1831 | 142 | |
AK294747.1 | - | 10 - 11 | 1918 | 142 | |
AK294777.1 | - | 10 - 11 | 1868 | 142 | |
AK299880.1 | - | 9 - 10 | 1861 | 142 | |
BC136453.1 | - | 9 - 10 | 2777 | 142 | |
BC136454.1 | - | 9 - 10 | 2777 | 142 |