Assay Design
Gene Transcripts
- Gene Symbol
- DNAJC19
- Entrez Gene ID
-
131118
- Gene Name
- DnaJ heat shock protein family (Hsp40) member C19
- Gene Aliases
-
PAM18, TIM14, TIMM14
- Chromosome Location
-
Chr.3: 180983709 - 180989774 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_033723.1 |
|
3 - 4 |
|
109 |
GenBank mRNA |
AW161751.1 |
- |
3 - 4 |
168 |
109 |
CB052107.1 |
- |
3 - 4 |
|
109 |
View Full Product Details