Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083885.2 | NP_001077354.2 | 3 - 4 | 457 | 71 |
NM_001173425.1 | NP_001166896.1 | 3 - 4 | 1621 | 71 | |
NM_015404.3 | NP_056219.3 | 3 - 4 | 1621 | 71 | |
XM_011518484.1 | XP_011516786.1 | 3 - 4 | 1674 | 71 | |
XM_011518485.1 | XP_011516787.1 | 4 - 5 | 1522 | 71 | |
XM_011518486.1 | XP_011516788.1 | 3 - 4 | 1674 | 71 | |
XM_011518488.1 | XP_011516790.1 | 3 - 4 | 1674 | 71 | |
XM_011518489.2 | XP_011516791.1 | 3 - 4 | 1674 | 71 | |
XM_011518491.2 | XP_011516793.1 | 3 - 4 | 1674 | 71 | |
XM_011518492.1 | XP_011516794.1 | 3 - 4 | 1674 | 71 | |
XM_011518494.2 | XP_011516796.1 | 3 - 4 | 1674 | 71 | |
GenBank mRNA | AB040959.1 | - | 3 - 4 | 1135 | 71 |
AL110228.1 | - | 3 - 4 | 415 | 71 | |
BC014524.1 | - | - | 2121 | 71 | |
BC136416.1 | - | 3 - 4 | 1370 | 71 | |
BC142684.1 | - | 3 - 4 | 1596 | 71 | |
DA502065.1 | - | 3 - 4 | 457 | 71 |