Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083885.2 | NP_001077354.2 | 8 - 9 | 1186 | 67 |
NM_001173425.1 | NP_001166896.1 | 8 - 9 | 2350 | 67 | |
NM_015404.3 | NP_056219.3 | 8 - 9 | 2350 | 67 | |
XM_011518484.1 | XP_011516786.1 | 9 - 10 | 2436 | 67 | |
XM_011518485.1 | XP_011516787.1 | 10 - 11 | 2284 | 67 | |
XM_011518486.1 | XP_011516788.1 | 9 - 10 | 2436 | 67 | |
XM_011518487.1 | XP_011516789.1 | 8 - 9 | 2310 | 67 | |
XM_011518488.1 | XP_011516790.1 | 7 - 8 | 2193 | 67 | |
XM_011518495.1 | XP_011516797.1 | 6 - 7 | 688 | 67 | |
GenBank mRNA | AB040959.1 | - | 8 - 9 | 1864 | 67 |
AK022854.1 | - | 4 - 5 | 1064 | 67 | |
AL110228.1 | - | 8 - 9 | 1144 | 67 | |
BC014524.1 | - | - | 2850 | 67 | |
BC136416.1 | - | 8 - 9 | 2099 | 67 | |
BC142614.1 | - | 8 - 9 | 2350 | 67 | |
BC142684.1 | - | 8 - 9 | 2325 | 67 |