Assay Design



Gene Transcripts

Gene Symbol
NIPAL4
Entrez Gene ID
348938
Gene Name
NIPA like domain containing 4
Gene Aliases
ARCI6, ICHTHYIN, ICHYN
Chromosome Location
Chr.5: 157460019 - 157474722 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099287.1 NP_001092757.1 1 - 2 341 68
NM_001172292.1 NP_001165763.1 1 - 2 341 68
GenBank mRNA AK026158.1 - 1 - 2 145 68
AK296972.1 - 1 - 2 341 68
AK308830.1 - 1 - 2 145 68
DB153698.1 - 1 - 2 292 68
DC348015.1 - 1 - 2 341 68

View Full Product Details