Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016604.3 | NP_057688.2 | 4 - 5 | 779 | 99 |
XM_005272018.4 | XP_005272075.1 | 4 - 5 | 638 | 99 | |
XM_011543488.2 | XP_011541790.1 | 4 - 5 | 494 | 99 | |
XM_011543489.2 | XP_011541791.1 | 4 - 5 | 953 | 99 | |
GenBank mRNA | AB029005.2 | - | 4 - 5 | 659 | 99 |
AF251039.1 | - | 3 - 4 | 389 | 100 | |
AF338242.1 | - | 4 - 5 | 779 | 99 | |
BC073869.1 | - | 3 - 4 | 281 | 99 | |
BC146788.1 | - | 4 - 5 | 660 | 99 | |
BG390435.1 | - | 3 - 4 | 280 | 99 |