Assay Design



Gene Transcripts

Gene Symbol
RALYL
Entrez Gene ID
138046
Gene Name
RALY RNA binding protein-like
Gene Aliases
HNRPCL3
Chromosome Location
Chr.8: 84182629 - 84921844 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100391.1 NP_001093861.1 4 - 5 540 123
NM_001100392.1 NP_001093862.1 5 - 6 783 123
NM_001100393.1 NP_001093863.1 4 - 5 751 123
NM_001287244.1 NP_001274173.1 3 - 4 245 123
NM_173848.5 NP_776247.3 4 - 5 1464 123
XM_017013059.1 XP_016868548.1 6 - 7 834 123
XM_017013060.1 XP_016868549.1 4 - 5 2984 123
XM_017013061.1 XP_016868550.1 4 - 5 2136 123
XM_017013062.1 XP_016868551.1 6 - 7 943 123
XM_017013063.1 XP_016868552.1 4 - 5 706 123
XM_017013064.1 XP_016868553.1 5 - 6 1057 123
XM_017013065.1 XP_016868554.1 4 - 5 605 123
XM_017013072.1 XP_016868561.1 4 - 5 692 123
XM_017013073.1 XP_016868562.1 4 - 5 692 123
XM_017013074.1 XP_016868563.1 4 - 5 605 123
XM_017013076.1 XP_016868565.1 4 - 5 485 123
XM_017013077.1 XP_016868566.1 5 - 6 1735 123
XM_017013078.1 XP_016868567.1 4 - 5 751 123
XM_017013079.1 XP_016868568.1 5 - 6 734 123
XM_017013083.1 XP_016868572.1 3 - 4 406 123
XM_017013085.1 XP_016868574.1 4 - 5 288 123
XM_017013086.1 XP_016868575.1 3 - 4 245 123
XM_017013087.1 XP_016868576.1 3 - 4 245 123
GenBank mRNA AK094543.1 - 4 - 5 695 123
AK095560.1 - 5 - 6 734 123
AK123437.1 - 3 - 4 245 123
BC031090.1 - 4 - 5 529 123

View Full Product Details