Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015473.3 | NP_056288.2 | 33 - 34 | 5642 | 140 |
GenBank mRNA | AB037737.1 | - | 24 - 25 | 4090 | 140 |
AK000343.1 | - | 11 - 12 | 1928 | 140 | |
BC045764.1 | - | 27 - 28 | 4655 | 140 | |
BC063867.1 | - | 15 - 16 | 2666 | 140 |