Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012244.3 | NP_036376.2 | 2 - 3 | 1104 | 89 |
GenBank mRNA | AB037669.1 | - | 2 - 3 | 1067 | 89 |
AF135828.1 | - | 2 - 3 | 579 | 89 | |
AF171669.1 | - | 2 - 3 | 581 | 89 | |
AK223610.1 | - | 2 - 3 | 1103 | 89 | |
AK313465.1 | - | 2 - 3 | 358 | 89 | |
AL365342.1 | - | 2 - 3 | 1089 | 89 | |
AL365343.2 | - | 2 - 3 | 608 | 89 | |
AL365414.1 | - | 2 - 3 | 608 | 89 | |
BC052250.1 | - | 2 - 3 | 387 | 89 | |
Y18483.1 | - | 2 - 3 | 1088 | 89 |