Assay Design



Gene Transcripts

Gene Symbol
P2RY2
Entrez Gene ID
5029
Gene Name
purinergic receptor P2Y2
Gene Aliases
HP2U, P2RU1, P2U, P2U1, P2UR, P2Y2, P2Y2R
Chromosome Location
Chr.11: 73200416 - 73246743 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005274019.4 XP_005274076.1 1 - 2 90 88

View Full Product Details