Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171137.1 | NP_001164608.1 | 1 - 2 | 141 | 69 |
NR_033234.1 | 1 - 2 | 129 | 69 | ||
XM_017014891.1 | XP_016870380.1 | 1 - 2 | 66 | 69 | |
GenBank mRNA | AK027890.1 | - | 1 - 2 | 183 | 69 |
AL136866.1 | - | 1 - 2 | 141 | 69 | |
AL833947.1 | - | 1 - 2 | 129 | 69 | |
AL834150.1 | - | 1 - 2 | 129 | 69 | |
BC108731.1 | - | 1 - 2 | 158 | 69 |